Learn More
Sino Biological Anti-SLAMF6 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11945MM1020
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Ly108 / SLAMF6 (rh Ly108 / SLAMF6; Catalog#11945-H08H; Q96DU3-1; Met1-Met226). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1420