Learn More
Sino Biological Recombinant Anti-SLAMF6 Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 11945R105A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Ly108/SLAMF6 (rh Ly108/SLAMF6; Catalog#11945-H08H; Q96DU3-1; Met1-Met226) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-240-9881