Learn More
Sino Biological Human SLAMF6 MAb 100TS (Sino Biological)
Supplier:  Sino Biological 11945R105P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Ly108/SLAMF6 (rh Ly108/SLAMF6; Catalog#11945-H08H; Q96DU3-1; Met1-Met226) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-237-2518