Learn More
Sino Biological Human SLAMF6 Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000786094
Encompass_Preferred
The Human SLAMF6 ELISA Pair Set is for the quantitative determination of Human SLAMF6.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human SLAMF6 coated on a 96-well plate. Standards and samples are added to the wells, and any Human SLAMF6 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human SLAMF6 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human SLAMF6 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86094 Supplier Sino Biological Supplier No. SEK1194515
The Human SLAMF6 ELISA Pair Set is for the quantitative determination of Human SLAMF6.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human SLAMF6 coated on a 96-well plate. Standards and samples are added to the wells, and any Human SLAMF6 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human SLAMF6 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human SLAMF6 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.