Learn More
Sino Biological Anti-SLAMF6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11945RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Ly108 / SLAMF6 (rh SLAMF6; Catalog#11945-H08H; Q96DU3-1; Met1-Met226). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-9873