Learn More
Sino Biological Recombinant Human SLAMF7/CRACC/CS1 Protein (hFc Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 11691H02HB20
Encompass_Preferred
A DNA sequence encoding the human SLAMF7 (NP_001269521.1) (Met1-Met226) was expressed with the Fc region of human IgG1 at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4611