Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human SLC17A1 (aa 39-79) Control Fragment Recombinant Protein

Catalog No. RP106288
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP106288 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP106288 Supplier Invitrogen™ Supplier No. RP106288
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (63%), Rat (63%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65625. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SLC17A1 (Solute Carrier Family 17 Member 1) is a Protein Coding gene. Diseases associated with SLC17A1 include Gout and Hyperuricemia. Among its related pathways are Transport of glucose and other sugars, bile salts and organic acids, metal ions and amine compounds and Uricosurics Pathway, Pharmacodynamics. Gene Ontology (GO) annotations related to this gene include symporter activity and phosphate ion transmembrane transporter activity. An important paralog of this gene is SLC17A2. Also, it is involved in cotransport of sodium and phosphate.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14916
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 6568
Name Human SLC17A1 (aa 39-79) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Na(+)/PI cotransporter 1; Na/Pi-4; Napi1; Napi-1; Npt1; NPT-1; OATV1; organic anion transporter OATv1; renal Na(+)-dependent phosphate cotransporter 1; Renal sodium-dependent phosphate transport protein 1; renal sodium-phosphate transport protein 1; Slc17a1; sodium phosphate transporter; sodium/phosphate cotransporter 1; sodium/phosphate type I cotransporter; sodium-dependent phosphate transport protein 1; sodium-dependent phosphate/anion cotransporter; solute carrier family 17 (organic anion transporter), member 1; solute carrier family 17 (sodium phosphate), member 1; solute carrier family 17 (sodium/hydrogen exchanger), member 1; solute carrier family 17 (vesicular glutamate transporter), member 1; solute carrier family 17 member 1; solute carrier family 17 vesicular glutamate transporter), member 1
Common Name SLC17A1
Gene Symbol Slc17a1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence CLNLTMVVMVNSTDPHGLPNTSTKKLLDNIKNPMYNWSPDI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.