Learn More
BIOHIPPO Human Slc26a5 Microsample
SDP
Catalog No. 5000313851
Encompass
Human Slc26a5 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of Slc26a5 in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 064 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human prestin Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human prestin Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human prestin biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13851 Supplier BIOHIPPO Supplier No. BHE15210588
Human Slc26a5 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of Slc26a5 in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 064 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human prestin Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human prestin Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human prestin biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color