Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC26A6 Partial ORF (NP_075062, 666 a.a. - 738 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7101912 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-962-226 25 μg
89-962-225 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-962-226 Supplier Abnova™ Supplier No. H00065010Q01L

Used for AP, Array, ELISA, WB-Re

This gene belongs to the solute carrier 26 family, whose members encode anion transporter proteins. This particular family member encodes a protein involved in transporting chloride, oxalate, sulfate and bicarbonate. Several alternatively spliced transcript variants of this gene, encoding distinct isoforms, have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]

Sequence: KSLKNIFHDFREIEVEVYMAACHSPVVSQLEAGHFFDASITKKHLFASVHDAVTFALQHPRPVPDSPVSVTRL

Specifications

Accession Number NP_075062
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 65010
Molecular Weight (g/mol) 33.77kDa
Name SLC26A6 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen KSLKNIFHDFREIEVEVYMAACHSPVVSQLEAGHFFDASITKKHLFASVHDAVTFALQHPRPVPDSPVSVTRL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp586E1422
Common Name SLC26A6
Gene Symbol SLC26A6
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.