Learn More
Bon Opus Biosciences Recombinant Human SLC39A6 (C-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP107550UG
Encompass_Preferred
Recombinant Human SLC39A6 (C-hFc) is produced by Human Cells. The target gene encoding Phe29-phe329 is expressed with a C-hFc tag. Accession# Q13433-1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1075
Catalog No. 50-217-2673