Learn More
Sino Biological Recombinant Human CD98 Protein (FITC conjugated, His Tag)
Catalog No. 502993112
Encompass_Preferred
A DNA sequence encoding the Human SLC3A2 (P08195) (Arg206-Ala630) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-299-3112 Supplier Sino Biological Supplier No. 12206H07HF200
A DNA sequence encoding the Human SLC3A2 (P08195) (Arg206-Ala630) was expressed, with a polyhistidine tag at the N-terminus.