Learn More
Sino Biological Human SLITL2/VASN Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785544
Encompass_Preferred
The Human Vasorin / VASN ELISA Pair Set is for the quantitative determination of Human Vasorin / VASN.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human Vasorin / VASN coated on a 96-well plate. Standards and samples are added to the wells, and any Human Vasorin / VASN present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human Vasorin / VASN monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human Vasorin / VASN present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85544 Supplier Sino Biological Supplier No. SEK138545
The Human Vasorin / VASN ELISA Pair Set is for the quantitative determination of Human Vasorin / VASN.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human Vasorin / VASN coated on a 96-well plate. Standards and samples are added to the wells, and any Human Vasorin / VASN present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human Vasorin / VASN monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human Vasorin / VASN present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.