Learn More
ACROBiosystems Biotinylated Human SLITRK6 protein, His,Avitag™
Catalog No. 502718918
Encompass_Preferred
SLIT and NTRK-like protein 6 (SLITRK6) belongs to the SLITRK family. SLITRK6 contains 11 LRR (leucine-rich) repeats, two LRRCT domains and two LRRNT domains. In adult brain, SLITRK6 is highly expressed in putamen and also is expressed in adult, fetal lung and fetal liver. SLITRK6 is also expressed at high levels in some brain tumors. Overexpression of SLITRK6 inhibites neurite outgrowth in cultured neurons.
Catalog No. 50-271-8918 Supplier ACROBiosystems Supplier No. SL6H82E3200UG
 Restricted Item
Add to Cart
Add to Cart
SLIT and NTRK-like protein 6 (SLITRK6) belongs to the SLITRK family. SLITRK6 contains 11 LRR (leucine-rich) repeats, two LRRCT domains and two LRRNT domains. In adult brain, SLITRK6 is highly expressed in putamen and also is expressed in adult, fetal lung and fetal liver. SLITRK6 is also expressed at high levels in some brain tumors. Overexpression of SLITRK6 inhibites neurite outgrowth in cultured neurons.