Learn More
Sino Biological Anti-SLITRK6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13922T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SLITRK6 (rh SLITRK6; Catalog#13922-H08H; Q9H5Y7; Met1-Ser608). SLITRK6 specific IgG was purified by Human SLITRK6 affinity chromatography.
Catalog No. 50-255-4157