Learn More
Raybiotech Inc Human SMAC ELISA Kit
Supplier:  Raybiotech Inc ELHSMAC1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 100 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detect human SMAC. Other species not determined yet.
Catalog No. 50-194-9736