Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Smad2 [pSpS465/467] ELISA™ Kit

Catalog No. 502545027
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-5027 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-5027 Supplier Invitrogen™ Supplier No. KHO8011
Only null left
Add to Cart
Add to Cart

ELISA

SMAD2 (MADH2, MAD2) regulates multiple cellular processes, such as cell proliferation, apoptosis, and differentiation. SMAD2 interacts with the TGF-beta receptors through its interaction with the SMAD anchor for receptor activation into the nucleus is a central event in TGF beta signaling. Phosphorylation of threonine 8 in the calmodulin-binding region of the MH1 domain by extracellular signal regulated kinase 1 (ERK1) enhances SMAD2 transcriptional activity, which is negatively regulated by calmodulin. In response to the TGF-beta signal, SMAD2 is phosphorylated by the TGF-beta receptors. The phosphorylation induces the dissociation of this protein with SARA and the association with the family member SMAD4. The association with SMAD4 is important for the translocation of this protein into the nucleus, where it binds to target promoters and forms a transcription repressor complex with other cofactors. SMAD2 can also be phosphorylated by activin type 1 receptor kinase, and mediates the signal from the activin. Alternatively spliced transcript variants encoding the SMAD2 protein have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15796
Assay Range 0 to 10 U/mL
Assay Sensitivity 0.42 U/mL
Conjugate HRP
Product Type ELISA
Sample Type Cell Lysates
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 4087
Gene Symbol SMAD2
Interassay CV 6.6%
Intraassay CV 3.9
Kit Contents Pre-coated 96 well plate, Standard (2 vials), Assay Buffer concentrate, Biotinylated Detection Antibody, Streptavidin-HRP, Wash Buffer, Substrate Solution, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 10 μL
Storage Requirements 2°C to 8°C
Target SMAD2
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 30 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.