Learn More
Sino Biological Anti-Smad5 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101483T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Smad5, and purified by antigen affinity chromatography.
Catalog No. 50-240-8434