Learn More
Sino Biological Human SMAD9 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 101177T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human SMAD9, and purified by antigen affinity chromatography.
Catalog No. 50-237-2099