Learn More
Sciencell Research Laboratories Human Small Airway Epithelial, Isolated from human lung tissue. HPSAEpiC are cryopreserved at passage one and delivered frozen. Each vial contains >5 x 105 cells in 1 ml volume.
Catalog No. NC2741778
Encompass
The small airways are located at the interface between alveoli and conducting airways. Airway epithelial cells form a continuous lining in the airways and play a unique role as a protective physical and functional barrier to external deleterious agents. Small airway epithelial cells (SAEpiC) regulate the immune responses by contributing to host defense through chemokine production and adhesion molecule expression. They also produce liquids contributing to pulmonary fluid balance. Many airway diseases, such as asthma, bronchiolitis, chronic obstructive pulmonary disease, and cystic fibrosis, involve damage to the airway surface epithelium. The study of human SAEpiC may help to identify new therapeutic options for preventing airway disorders.HPSAEpiC from ScienCell Research Laboratories are isolated from human lung tissue. HPSAEpiC are cryopreserved at passage one and delivered frozen. Each vial contains >5 x 105 cells in 1 ml volume. H
Catalog No. NC2741778 Supplier Sciencell Research Laboratories Supplier No. 3230
The small airways are located at the interface between alveoli and conducting airways. Airway epithelial cells form a continuous lining in the airways and play a unique role as a protective physical and functional barrier to external deleterious agents. Small airway epithelial cells (SAEpiC) regulate the immune responses by contributing to host defense through chemokine production and adhesion molecule expression. They also produce liquids contributing to pulmonary fluid balance. Many airway diseases, such as asthma, bronchiolitis, chronic obstructive pulmonary disease, and cystic fibrosis, involve damage to the airway surface epithelium. The study of human SAEpiC may help to identify new therapeutic options for preventing airway disorders.HPSAEpiC from ScienCell Research Laboratories are isolated from human lung tissue. HPSAEpiC are cryopreserved at passage one and delivered frozen. Each vial contains >5 x 105 cells in 1 ml volume. H