Learn More
Biomatik Corporation Human Small Proline Rich Protein 4 (SPRR4) ELISA Kit
Supplier:  Biomatik Corporation EKU11433
Encompass_Preferred
Human Small Proline Rich Protein 4 (SPRR4) ELISA Kit for detecting SPRR4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96PI1. Gene ID: 163778. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7651