Learn More
Bioworld HUMAN SMALL UBIQUITIN-RELAT
Supplier:  Bioworld 220604911
Encompass_Preferred
HUMAN SMALL UBIQUITIN-RELATED MODIFIER 1 (RECOMBINANT): Recombinant Human SUMO-I/Sentrin-I Source: E.Coli Background: Human SUMO-1 (small ubiquitin-like modifier) is an 11-kDa protein that shares 18% sequence homology with ubiquitin....
Catalog No. 50-253-586