Learn More
Biomatik Corporation Human Small Ubiquitin Related Modifier Protein 2 (SUMO2) ELISA Kit
Supplier:  Biomatik Corporation EKU07377
Encompass_Preferred
Human Small Ubiquitin Related Modifier Protein 2 (SUMO2) ELISA Kit for detecting SUMO2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.126ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P61956. Gene ID: 6613. Target Synonyms: HSMT3; SMT3B; SMT3H2; SUMO-3; Sentrin-2; Ubiquitin-like protein SMT3B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7558