Learn More
Sino Biological Anti-BRG1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100615T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human SMARCA4 / BRG1, and purified by antigen affinity chromatography.
Catalog No. 50-241-1613