Learn More
Sino Biological Human SMIM4 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 12724RP01100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human C3orf78 / SMIM4 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-237-4437