Learn More
Biomatik Corporation Human Smoothened (SMO) ELISA Kit
Supplier:  Biomatik Corporation EKU11726
Encompass_Preferred
Human Smoothened (SMO) ELISA Kit for detecting SMO in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99835. Gene ID: 6608. Target Synonyms: Gx; SMOH. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4676