Learn More
Sino Biological Human SMPDL3A PAb 100ul (Sino Biological)
Supplier:  Sino Biological 13559T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SMPDL3A (rh SMPDL3A; Catalog#13559-H08H; Q92484; Met1-Tyr453). SMPDL3A specific IgG was purified by Human SMPDL3A affinity chromatography.
Catalog No. 50-237-2902