Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human SMU1 (aa 2-106) Control Fragment Recombinant Protein

Catalog No. RP92934
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP92934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP92934 Supplier Invitrogen™ Supplier No. RP92934
  • $283.00
    $212.25 / Each of 1

    Save $70.75

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54062. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SMU1 (suppressor of Mec8 and Unc-52 homolog), also known as BWD (brain-enriched WD), is a member of the WD-repeat SMU1 family and contains one CTLH domain, one LisH domain and six WD-repeats. SMU1 is a homolog of the C. elegans protein Smu1, a ubiquitously expressed nuclear protein that is believed to play a role in alternative splicing events of Unc-52. In mammals, SMU1 is a component of the spliceosome and appears to exhibit a conserved function, affecting the splicing of the mammalian Unc-52 homolog, namely Perlecan. Mutations in the gene encoding SMU1 may affect the function of the spliceosome.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q2TAY7
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 55234
Name Human SMU1 (aa 2-106) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2600001O03Rik; 2610203K23Rik; AB044414; AI845086; AW556129; Brain-enriched WD repeat-containing protein; brain-enriched WD-repeat protein; BWD; DNA replication regulator and spliceosomal factor; fSAP57; functional spliceosome-associated protein 57; homolog of C. elegans smu-1; SMU1; SMU-1; smu-1 suppressor of mec-8 and unc-52 homolog; smu-1 suppressor of mec-8 and unc-52 homolog (C. elegans); smu-1 suppressor of mec-8 and unc-52 protein homolog; suppressor of Mec and Unc defects 1 homolog; WD40 repeat-containing protein SMU1; WD40 repeat-containing protein SMU1, N-terminally processed
Common Name SMU1
Gene Symbol Smu1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SIEIESSDVIRLIMQYLKENSLHRALATLQEETTVSLNTVDSIESFVADINSGHWDTVLQAIQSLKLPDKTLIDLYEQVVLELIELRELGAARSLLRQTDPMIML
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.