Learn More
Sino Biological Recombinant Human SMYD3 Protein (His & FLAG Tag)
Supplier:  Sino Biological 11230H18B50
Encompass_Preferred
A DNA sequence encoding the Human SMYD3 (NP_001161212.1) (Met1-Ser428) was expressed with the flag tag at the N-terminus and the histidine tag at the C-terminus.
Catalog No. 50-240-8671