Learn More
Biomatik Corporation Human SNAP Associated Protein (SNAPAP) ELISA Kit
Supplier:  Biomatik Corporation EKU07381
Encompass_Preferred
Human SNAP Associated Protein (SNAPAP) ELISA Kit for detecting SNAPAP in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.066ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O95295. Gene ID: 23557. Target Synonyms: SNAPIN; SNAP-25-Binding Protein; Biogenesis of lysosome-related organelles complex 1 subunit 7; Synaptosomal-associated protein 25-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7337