Learn More
BIOHIPPO Human SNAP23 Microsample
Catalog No. 5000314809
Encompass
Human SNAP23 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SNAP23 in serum plasma and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 27 2 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human SNAP23 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human SNAP23 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human SNAP23 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14809 Supplier BIOHIPPO Supplier No. BHE15210600
Human SNAP23 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SNAP23 in serum plasma and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 27 2 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human SNAP23 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human SNAP23 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human SNAP23 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color