Learn More
Sino Biological Recombinant Anti-alpha-Synuclein Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 12093R002100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human alpha-Synuclein (rh alpha-Synuclein; Catalog#12093-HNAE; P37840-1; Met1-Ala140).
Catalog No. 50-241-2327