Learn More
Sino Biological Human SNCA PAb 100ul (Sino Biological)
Supplier:  Sino Biological 69009A08H50
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human alpha-Synuclein / SNCA (rh alpha-Synuclein / SNCA; Catalog#12093-HNAE; P37840-1; Met1-Ala140). alpha-Synuclein / SNCA specific IgG was purified by Human alpha-Synuclein / SNCA affinity chromatography.
Catalog No. 50-237-4356