Learn More
Sino Biological Recombinant Human alpha-Synuclein Protein (His & AVI Tag), Biotinylated
Supplier:  Sino Biological 12093H49EB100
Encompass_Preferred
A DNA sequence encoding the Human SNCA (P37840-1) (Met1-Ala140) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-4460