Learn More
Biomatik Corporation Human SNCA (Alpha-synuclein) ELISA Kit
Supplier:  Biomatik Corporation EKF58419
Encompass_Preferred
Human SNCA (Alpha-synuclein) ELISA Kit for detecting SNCA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P37840. Target Synonyms: SNCA; NACP; PARK1; PARK4; PD1; SNCA; Lewy body 4; non A-beta component of AD amyloid; Non-A beta component of AD amyloid; non-A4 component of amyloid; Non-A4 component of amyloid precursor; PARK1; PARK4; synuclein; alpha; non A4 component of amyloid precursor; Synuclein-alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6685