Learn More
Sino Biological Human SNX2 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 200997T10100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human SNX2 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-237-3038