Learn More
Sino Biological Human SNX6 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 14483T48100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human SNX6, and purified by antigen affinity chromatography.
Catalog No. 50-237-4047