Learn More
Sino Biological Human SOD1 ELISA Pair Set 5PP (Sino Biological)
Catalog No. 502371758
Encompass_Preferred
Each vial contains 180 ng of recombinant SOD1. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 4 ng/mL is recommended.
Catalog No. 50-237-1758 Supplier Sino Biological Supplier No. SEK117275
Each vial contains 180 ng of recombinant SOD1. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 4 ng/mL is recommended.