Learn More
Sino Biological Anti-SOD1/Cu-Zn SOD Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 11727MM11F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SOD1 / Superoxide Dismutase (rh SOD1 / Superoxide Dismutase; Catalog#11727-H07E; NP_000445.1; Ala2-Gln154) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-240-8677