Learn More
Sino Biological Human SOD1 MAb 25TS (Sino Biological)
Supplier:  Sino Biological 11727R106F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human SOD1 / Superoxide Dismutase (rh SOD1 / Superoxide Dismutase; Catalog#11727-H07E; NP_000445.1; Ala2-Gln154) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-237-1162