Learn More
BIOHIPPO Human SOD1 Microsample
SDP
Catalog No. 5000315428
Encompass
Human SOD1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SOD1 in serum plasma tissue homogenates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 26 4 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human SOD1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human SOD1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human SOD1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15428 Supplier BIOHIPPO Supplier No. BHE15211933
Human SOD1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SOD1 in serum plasma tissue homogenates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 26 4 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human SOD1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human SOD1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human SOD1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color