Learn More
Sino Biological Anti-SOD1/Cu-Zn SOD Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 201220T36100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human SOD1 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-240-9943