Learn More
BIOHIPPO Human SOD3 Microsample
SDP
Catalog No. 5000315384
Encompass
Human SOD3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SOD3 in serum plasma cell lysates and other biological fluids It provides a detection range of 125-8000 pg/mL with a sensitivity of 55 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human SOD3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human SOD3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human SOD3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15384 Supplier BIOHIPPO Supplier No. BHE15211898
Human SOD3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SOD3 in serum plasma cell lysates and other biological fluids It provides a detection range of 125-8000 pg/mL with a sensitivity of 55 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human SOD3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human SOD3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human SOD3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color