Learn More
IBL America Alpha Klotho (α - Klotho) Human Soluble ELISA
Supplier:  IBL America 27998
Encompass_Preferred
Solid phase sandwich ELISA for the determination of human soluble Alpha Klotho ( α-Klotho ) in serum and EDTA-plasma. For research use only, not for use in diagnostic procedures.
Catalog No. 50-212-0382