Learn More
American Research Products Inc Human Soluble Cluster of Diffe
Catalog No. 5000330134
Encompass
Human Soluble Cluster of Differentiation 80 (sCD80) ELISA Kit from CUSABIO Detection Range 125 pg/mL-8000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma cell culture supernates Method Sandwich
Catalog No. 50-003-30134 Supplier American Research Products Inc Supplier No. CSBE15768H
Human Soluble Cluster of Differentiation 80 (sCD80) ELISA Kit from CUSABIO Detection Range 125 pg/mL-8000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma cell culture supernates Method Sandwich