Learn More
Abcam Human soluble EGFR ELISA Kit, 1 x 96 tests, Sandwich ELISA, Assay Duration 90 min, Single wash, Sensitivity 36.63 pg/mL, Colorimetric detection at 450 nm
Supplier:  Abcam AB2695581X96TESTS
Encompass_Preferred
Human soluble EGFR ELISA Kit, 1 x 96 tests, Sandwich ELISA, Assay Duration 90 min, Single wash, Sensitivity 36.63 pg/mL, Colorimetric detection at 450 nm
Catalog No. NC2382173