Learn More
American Research Products Inc Human soluble interleukin 2 re
Catalog No. 5000326086
Encompass
Human soluble interleukin-2 receptor IL-2sRa ELISA kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26086 Supplier American Research Products Inc Supplier No. CSBE04629H
Human soluble interleukin-2 receptor IL-2sRa ELISA kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method Sandwich