Learn More
Description
Includes
- Assay Diluent
- Capture Bead Diluent
- Detection Reagent Diluent
- Capture Bead Diluent for Serum/Plasma
- Wash Buffer
- Instrument Setup Bead A1
- Instrument Setup Bead A9
- Instrument Setup Bead F1
- Instrument Setup Bead F9
- PE Instrument Setup Bead F1
- PE Positive Control Detector
Buffers and instrument setup reagents provided in this kit have been optimized for analysis of analytes in tissue culture supernatants, plasma, and serum samples.
- Provides sufficient reagents for the quantitative analysis of 100 samples and 10 instrument setup procedures or 500 samples and 10 instrument setup procedures
- Has been optimized for use with the CBA Human Soluble Protein Flex Sets
- Working assay range for most analytes in this system is 10 to 2,500pg/mL
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| No. of Tests | 500 |
| Includes | 1 bottle 150mL assay diluent, 1 bottle 30mL capture bead diluent, 1 bottle 30mL detection reagent diluent, 1 bottle 30mL capture bead diluent for Serum/Plasma, 1 bottle 650mL wash buffer, 1 vial 0.25 mL instrument setup bead A1, 1 vial 0.25mL instrument |
| Validated Application | RUO |
| Product Type | Cytometric Bead Array (CBA) Assay |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.