Learn More
Biomatik Corporation Human Soluble Protein-100B, S-100B Elisa Kit
Supplier:  Biomatik Corporation EKC31323
Encompass_Preferred
Human Soluble Protein-100B, S-100B Elisa Kit for detecting S100 Calcium Binding Protein B in serum, plasma, tissue homogenates, cerebrospinal fluid (csf). Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04271; S100B. Target Synonyms: NEF; Protein S100 B; Protein S100-B; S 100 calcium binding protein beta chain; S 100 protein beta chain; S-100 protein beta chain; S-100 protein subunit beta; S100; S100 calcium binding protein beta (neural); S100 calcium-binding protein B; S100 protein beta chain; S100B; S100B_HUMAN; S100beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5455