Learn More
American Research Products Inc Human soluble receptor actiVat
Catalog No. 5000329925
Encompass
Human soluble receptor actiVator of nuclear factor-kB ligand sRANKL ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma cell culture supeernates tissue homogenates Method Sandwich
Catalog No. 50-003-29925 Supplier American Research Products Inc Supplier No. CSBE05125H
Human soluble receptor actiVator of nuclear factor-kB ligand sRANKL ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma cell culture supeernates tissue homogenates Method Sandwich