Learn More
Biomatik Corporation Human Solute Carrier Family 16, Member 3 (SLC16A3) ELISA Kit
Supplier:  Biomatik Corporation EKU07393
Encompass_Preferred
Human Solute Carrier Family 16, Member 3 (SLC16A3) ELISA Kit for detecting SLC16A3 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.28ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O15427. Gene ID: 9123. Target Synonyms: MCT4; Monocarboxylic Acid Transporter 4; Monocarboxylate Transporter 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8755